Skip to Content
MilliporeSigma
All Photos(1)

Documents

SUP-T1

95013123, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SUP-T1, 95013123

biological source

human blood

growth mode

Suspension

karyotype

92XXXY, -2, -2, -9, -9, 2inv(2*)(p21q11), +2inv(2), del(2*)(Q21), 2del(4)(q31q35), 2del (5)(p13p14), 2del(6)(q23q27), inv(14)(q11q32), t(7;9)(q34,q34), +der(9)t(7;9). Undergone Tetraploidisation

morphology

Lymphoblast

products

Not specified

receptors

CD4

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T cell lymphoblastic Lymphoma

Cell Line Description

Derived from a Malignant pleural effusion taken from an eight year old caucasian male with T cell lymphoblastic lymphoma. These cells are E-rosette, Leu 5, OKT9 and CALLA negative. They also express β2M, Leu 1, 2, 3a, 4, 6, 9 and OKT10 antigens and are CD4 positive. These cells are negative for Tac, HLA-DR and surface immunoglobulin.

Application

Cells have been used for growth of HIV-1
SUP-T1 has been used in a study targeting B-cell neoplasia with T-cell receptors recognizing a cluster of differentiation 20 (CD20)-derived peptide on patient-specific human leucocyte antigen (HLA). Existing appl-SUP-T1 cells have been used for growth of HIV-1.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2x100,000 to 2x1,000,000 cells/ml 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service