Skip to Content
MilliporeSigma
All Photos(1)

Documents

Ca Ski

87020501, human cervix (small bowel metastasis), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Ca Ski, 87020501

biological source

human cervix (small bowel metastasis)

growth mode

Adherent

karyotype

Modal no. 78

morphology

Epithelial

products

Beta Human chorionic gonadotropin (beta-hCG)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian cervical epidermoid carcinoma

Cell Line Description

Derived from an epidermoid carcinoma of the cervix metastatic to the small bowel mesentery of a 40 year old Caucasian. Ca Ski cells secrete the β subunit of human chorionic gonadotropin (β-h CG), express tumour-associated antigen and G6PD type B. Ca Ski cells have been reported to contain an integrated human papilloma virus 16 genome (HPV-16 about 600 copies per cell) and HPV-18 related sequences.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 8,12
D16S539: 11
D5S818: 13
D7S820: 8,11
THO1: 7
TPOX: 8
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service