Skip to Content
MilliporeSigma
All Photos(1)

Documents

CFPAC-1

91112501, human pancreas (liver metastasis), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CFPAC-1, 91112501

biological source

human pancreas (liver metastasis)

growth mode

Adherent

karyotype

Hyperdiploid, (65-75 chromosomes)

morphology

Epithelial

products

Not specified

receptors

Cystic fibrosis trans-membrane regulator (CFTR)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian pancreatic adenocarcinoma

Cell Line Description

CFPAC-1 is a ductal pancreatic adenocarcinoma derived by differential trypsinisation of explant cultures from a metastatic lesion in the liver of a 26 year old caucasian male with cystic fibrosis (CF). The cell line expresses cystic fibrosis trans-membrane regulator (CFTR) and ion transport properties consistent with the CF defect. CFPAC-1 cells show an epithelial morphology and polarisation with apical microvilli. Expression of cytokeratin and oncofoetal antigens characteristic of pancreatic duct cells have been described. The cells express the CF gene and show the most common CF mutation, deletion of three nucleotides result in a phenylalanine-508 deletion. CFPAC-1 cells are also tumorigenic in nude mice.

Application

Cystic fibrosis studies, genetic analysis

Culture Medium

IDMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service