Skip to Content
MilliporeSigma
All Photos(1)

Documents

11012002

Sigma-Aldrich

KYAE-1 cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human esophagus

Quality Level

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

P53 mutation c.455C>T

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Distal oesophageal adenocarcinoma,

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11,12
D16S539: 11
D5S818: 10,13
D7S820: 11,12
THO1: 6
TPOX: 8
vWA: 14

Culture Medium

RPMI-1640:Hams F12 1:1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 3 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. There is a requirement to change the culture medium every 2-3 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donna E Hansel et al.
The American journal of surgical pathology, 29(3), 390-399 (2005-02-24)
Esophageal adenocarcinoma arises through well-defined precursor lesions (Barrett esophagus), although only a subset of these lesions advances to invasive adenocarcinoma. The lack of markers predicting progression in Barrett esophagus, typical presentation at advanced stage, and limitations of conventional chemotherapy result
T Kan et al.
Biochemical and biophysical research communications, 286(4), 792-801 (2001-08-25)
Human esophageal cancer cell lines and human esophageal cancer tissues were profiled on cDNA microarrays. In esophageal cancer cell lines, KYAE and OE-33 (adenocarcinomas) were distinguished from KYSE series (squamous cell carcinomas). Although SK-GT-4 and TE7 were derived from adenocarcinomas
Jurjen J Boonstra et al.
Journal of the National Cancer Institute, 102(4), 271-274 (2010-01-16)
For decades, hundreds of different human tumor type-specific cell lines have been used in experimental cancer research as models for their respective tumors. The veracity of experimental results for a specific tumor type relies on the correct derivation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service