Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OE33 Cell Line human

96070808, human esophagus, Epithelial

Synonym(s):

JROECL33, JROECL33 Cells, OE-33 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

OE33 Cell Line human, 96070808, Caucasian esophageal carcinoma

biological source

human esophagus

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial

products

Not specified

receptors

MHC class I, ICAM-1

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian oesophageal carcinoma

Cell Line Description

The cell line OE33, also known as JROECL33, was established from the adenocarcinoma of the lower oesophagus (Barrett′s metaplasia) of a 73 year old female patient. The tumour was identified as pathological stage IIA (UICC) and showed poor differentiation. OE33 express HLA-A, -B and -C antigens (MHC class I) and ICAM-1 constitutively. Expression of HLA-DR (MHC class II) can be induced by treatment with interferon-gamma. The cells express epithelial cytokeratins and are tumourigenic in nude mice.

Application

OE33 cell line has been used to study the effect of microRNA-MiR-193b on autophagy and cell death on these cells. It has also been used to study the sensitivity of the cells to anti-c-Met monoclonal antibody ABT-700.
Study of oesophageal cancer

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum FBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 i.e. seeding at 1x10,000 cells/cm2 using 0.25% Trypsin or Trypsin/EDTA; 5% CO2; 37°C. Initially these cells grow slowly and can take up to 7 days until ready for the next split, 50% media changes will be necessary every 2-3 days (i.e. replacing half the old medium with fresh).

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service