Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106514
Biological source:
human skin
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Not specified
Morphology:
Not specified
biological source
human skin
packaging
tube of 5 μg 96020925-DNA-5UG, pkg of vial of cells 96020925-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Biochem/physiol Actions
Human Amelanotic Melanoma
This cell line has been derived by the selection of the human amelanotic melanoma cell line C32 for 6-thioguanine resistance. The parent cell line C32 was established from a melanoma from a 53 year old Caucasian male. The cells are resistant to 6-thioguanine and sensitive to HAT medium. They are deficient in hypoxanthine ? guanine phosphoribosyl transferase (HGPRT-). The cell line is also known as C32-r16TG. The cells are tumorigenic in nude mice inoculated subcutaneously with 10E7 cells.
Preparation Note
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 0.050mM 6-thioguanine + 10% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80 %) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 37 C; 5% CO2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.