Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A431 Cell Line human

85090402, human skin, Epithelial

Synonym(s):

A-431 Cells, A431/P Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

A431 Cell Line human, squamous carcinoma, 85090402

biological source

human skin

growth mode

Adherent

karyotype

Hypertriploid

morphology

Epithelial

products

Not specified

receptors

Epidermal growth factor (EGF)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human squamous carcinoma

Cell Line Description

Derived from an epidermal carcinoma of the vulva taken from an 85 yr old female. The cells carry large numbers of EGF binding sites and is an indicator cell for anti-TGF binding.

Application

A431 Cell Line human has been used:
  • to demonstrate that p38 mitogen-activated protein kinase enhances epidermal growth factor-induced apoptosis in A431 cells
  • to study melanoma-specific CD44 (a tumor marker and transmembrane receptor) alternative splicing pattern in A431 cells
  • to study the anti-cancer activity of antioxidant-rich Cameroonian medicinal plants
Cell cycle studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application note on how the CellASIC® ONIX2 microfluidic system can be used to analyze caspase-3 mediated apoptosis/cell death and cellular hypoxia in live immune and cancer cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service