Skip to Content
MilliporeSigma
All Photos(1)

Documents

11012009

ESO26

Synonym(s):

ESO-26 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human esophagus

Quality Level

growth mode

Suspension

karyotype

Not specified

morphology

Large spherical clumps of cells loosely adherent to the plastic

products

P53 mutation c.742C>T

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Adenocarcinoma of the gastroesophageal junction

Cell Line Description

ESO26 was established from a primary tumour located in the gastroesophageal junction and distal oesophagus of a 56 year-old Caucasian male in 2000 involving lymph nodes and distant metastases. ESO26 was found to be tumourigenic in female NMRI nude mice or NOD-SCID mice.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 9,10
D13S317: 9,11
D16S539: 9
D5S818: 12
D7S820: 11
THO1: 9
TPOX: 7,9
vWA: 14,18

Culture Medium

RPMI-1640 + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-9 x 105 cells/cm3. Cells are loosely adherent and can be dislodged by gentle tapping or use of 0.02% EDTA solution. 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service