Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCC203

Sigma-Aldrich

IGROV-1 Human Ovarian Cancer Cell Line

IGROV-1 human ovarian carcinoma cell line is a versatile model for ovarian carcinoma and investigation of drug resistance.

Synonym(s):

IGR-OV1, OV1/P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

application(s)

cell analysis

General description

The IGROV-1 cell line is well-established as a model for drug-resistant ovarian carcinoma. IGROV-1 cells lack hormone receptors and exhibit low to moderate sensitivity to several common chemotherapeutic agents, while remaining sensitive to cisplatin (1). IGROV-1 cells harbor a paracentric inversion of chromosome 3 and a translocation between chromosomes 2 and 5 (1). The IGROV-1 cell line exhibits hypermutation and has an overall mutation profile similar to endometrioid carcinoma (2). IGROV-1 cells appear in culture as confluent monolayers and are highly tumorigenic in vivo, forming solid tumors within 15 days in nude mice (1). IGROV-1 cells are characterized by expression of vitronectin and αvα3 integrin, essential for the observed migratory behavior of these cells toward ECM proteins (3), and express the cancer-associated target MDM2 (4).

Source:
IGROV-1 cells were established from a stage III ovarian solid primary tumor of a 47-year-old female patient (1).

Research Category:
Cancer

Cell Line Origin

Human, Cancer Cells

Packaging

≥1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service