Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5127

Sigma-Aldrich

Starch from wheat

Unmodified

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

wheat

Quality Level

form

solid

impurities

≤0.3% protein content

color

white

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

May be used to create starch-based scaffolds for studies of cells in 3-D culture applications.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Majid Haddad Momeni et al.
Biotechnology for biofuels, 13, 135-135 (2020-08-11)
Lytic polysaccharide monooxygenases (LPMOs) are often studied in simple models involving activity measurements of a single LPMO or a blend thereof with hydrolytic enzymes towards an insoluble substrate. However, the contribution of LPMOs to polysaccharide breakdown in complex cocktails of
Fang Xie et al.
International journal of biological macromolecules, 126, 525-530 (2018-12-28)
Starch-containing food normally has complex components and structure, which control its digestion kinetic properties and nutritional value. However, interactions among multiple food components and their effects on starch digestion kinetics are rarely reported. Here, we evaluated the influence of tea
Eiji Yamazaki et al.
Carbohydrate polymers, 94(1), 555-560 (2013-04-03)
Effect of the polysaccharide from leaves of Corchorus olitorius L. (PLC) on the freeze-thaw (FT) stability of corn starch gel was studied. PLC was incorporated into the starch gel at 0.7% and total solid was adjusted to 6.0%. The syneresis
Bruno R Cruz et al.
Carbohydrate polymers, 94(1), 594-602 (2013-04-03)
Starch isolation methods can change their physico-chemical and functional characteristics hindering the establishment of a starch-food functionality relation. A simple high yield and soft isolation method was applied for chestnut (Castanea sativa Mill) starch consisting in steeping and fruit disintegration
Daniëlle Haenen et al.
The Journal of nutrition, 143(12), 1889-1898 (2013-10-18)
Consumption of resistant starch (RS) has been associated with various intestinal health benefits, but knowledge of its effects on global gene expression in the colon is limited. The main objective of the current study was to identify genes affected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service