Skip to Content
MilliporeSigma

TDFC10

Dietary Fiber, Total, Assay Control Kit

sufficient for ≥10 assays

Synonym(s):

Total Dietary Fiber Controls Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12164500
EC Number:
232-679-6
NACRES:
NA.84
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for ≥10 assays

Quality Segment

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Other Notes

Standards for use as internal controls in conjunction with the Total Dietary Fiber Assay Kit (TDF100A)


Kit Components Only

Product No.
Description

  • Arabinogalactan (stractan) 1 g

  • Casein 5 g

  • β-Glucan 1 g

  • Pectin 1 g

  • Starch, corn 10 g

  • Starch, wheat 10 g

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library