Skip to Content
MilliporeSigma

1.11685

Starch (from wheat)

for biochemistry

Synonym(s):

Starch (from wheat), starch native from wheat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
EC Index Number:
232-679-6
NACRES:
NA.79
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

autoignition temp.

400 °C

pH

4-7 (25 °C, 100 g/L in H2O, slurry)

bulk density

550‑700 kg/m3

storage temp.

no temp limit

Analysis Note

Reducing sugars (as maltose): ≤ 0.5 %
Heavy metals (as Pb): ≤ 0.001 %
Sulfated ash: ≤ 0.6 %
Loss on drying: ≤ 13 %


Still not finding the right product?

Explore all of our products under Starch (from wheat)


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library