Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10120

Sigma-Aldrich

Amylopectin from maize

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

maize (





)

form

powder

ign. residue

≤0.1%

color

white to off-white

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O[C@H]5O[C@@H]([C@H]([C@@H]([C@H]5O)O)O)CO)CO)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C30H52O26/c31-1-6-11(35)13(37)19(43)28(50-6)55-24-9(4-34)52-27(21(45)16(24)40)48-5-10-25(56-29-20(44)14(38)12(36)7(2-32)51-29)17(41)22(46)30(53-10)54-23-8(3-33)49-26(47)18(42)15(23)39/h6-47H,1-5H2/t6-,7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26+,27+,28-,29-,30-/m1/s1

InChI key

WMGFVAGNIYUEEP-WUYNJSITSA-N

General description

α-1,4-glucan with α-1,6-cross-linking; one terminal group per 25 glucose units

Application

Amylopectin is a glucose-based, highly-branched polysaccharide which together with amylose makes starch. The glucosidic bonds of amylopectin are α-1,4 in the glucan chain and α-1,6 at the branch points. Amylopectin from maize differs in structure form amylopectin from other plant species and is used in the study of the development of starches.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Structural characterization of amylopectin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Baba et al.
Agricultural and Biological Chemistry, 48, 1763-1763 (1984)
David A Johnston et al.
Carbohydrate research, 346(17), 2777-2784 (2011-11-08)
Seven different starches from potato, rice, maize, waxymaize, amylomaize-VII, shoti, and tapioca, and potato amylose and potato amylopectin have been reacted with benzaldehyde, catalyzed by ZnCl(2), to give new water-soluble starches and water soluble-amylose and soluble-amylopectin. In contrast to the
Shazia Juna et al.
Journal of chromatography. A, 1219, 161-172 (2011-12-07)
A detailed study of the elution behaviour, apparent molecular characteristics and hydrodynamic properties of amylopectin-type fraction (isolated from normal corn starch) in aqueous media employing asymmetrical flow field-flow fractionation (AF4) was undertaken by systematically varying the channel flow (F(ch)), cross
Jheng-Hua Lin et al.
Carbohydrate polymers, 92(2), 1858-1864 (2013-02-13)
A set of 13 waxy rice genotypes prepared by chemically-induced mutation of non-waxy rice variety TNG67 and 7 waxy rice varieties widely planted in Taiwan were screened for various structural and gelatinization properties of starches. Wide variation on physicochemical properties
Varatharajan Vamadevan et al.
Carbohydrate polymers, 92(2), 1653-1659 (2013-02-13)
The relationship between the internal structure of amylopectin from diverse plants and thermal properties of the starch granules has been investigated. Correlations were found between structural parameters, such as number of building blocks in clusters, interblock chain length and length

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service