Skip to Content
MilliporeSigma

P9135

Pectin from citrus peel

Galacturonic acid ≥74.0 % (dried basis)

Synonym(s):

Poly-D-galacturonic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
232-553-0
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

citrus (peels)

Quality Segment

form

powder

composition

Galacturonic acid, ≥74.0% (dried basis)

impurities

≤10% Moisture

color

light brown

application(s)

advanced drug delivery

storage temp.

room temp

General description

Pectin is a polysaccharide is rich in galacturonic acid and possesses a complex structure and function.

Application

Pectin from citrus peel has been used:
  • in the comparative study of physicochemical, rheological and gelling properties of pectin among different extraction method
  • in starch suspension for measuring in vitro starch digestibility
  • to measure pectinase activity
  • in the production of pectinase on solid medium

Biochem/physiol Actions

Pectin is essential for plant development including growth, morphological changes and immunity. It serves as a stabilizing polymer and is useful in food and speciality products.
Pectin, a heterosaccharide component of terrestrial plant cell walls, is used as a substrate to identify, differentiate and characterized pectinase(s). Pectin is used to study its degradation by pectinolytic bacteria.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library