Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7260

Sigma-Aldrich

Starch from rice

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

rice

Quality Level

form

solid

impurities

≤1% protein content

color

white

pH

5.5-7.0

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1

InChI key

GUBGYTABKSRVRQ-ASMJPISFSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chonthira Sarawong et al.
Food chemistry, 143, 33-39 (2013-09-24)
Green banana flour was extruded through a co-rotating twin-screw extruder with constant barrel temperature. The objectives of this study were to determine the effect of extrusion cooking variables (feed moisture, FM, 20% and 50%; screw speed, SS, 200 and 400rpm)
Eiji Yamazaki et al.
Carbohydrate polymers, 94(1), 555-560 (2013-04-03)
Effect of the polysaccharide from leaves of Corchorus olitorius L. (PLC) on the freeze-thaw (FT) stability of corn starch gel was studied. PLC was incorporated into the starch gel at 0.7% and total solid was adjusted to 6.0%. The syneresis
Li Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5380-5385 (2013-03-20)
Three grinding stones from Shizitan Locality 14 (ca. 23,000-19,500 calendar years before present) in the middle Yellow River region were subjected to usewear and residue analyses to investigate human adaptation during the last glacial maximum (LGM) period, when resources were
Vikas Kumar et al.
Critical reviews in food science and nutrition, 52(10), 899-935 (2012-07-04)
Nonstarch polysaccharides (NSPs) occur naturally in many foods. The physiochemical and biological properties of these compounds correspond to dietary fiber. Nonstarch polysaccharides show various physiological effects in the small and large intestine and therefore have important health implications for humans.
Bruno R Cruz et al.
Carbohydrate polymers, 94(1), 594-602 (2013-04-03)
Starch isolation methods can change their physico-chemical and functional characteristics hindering the establishment of a starch-food functionality relation. A simple high yield and soft isolation method was applied for chestnut (Castanea sativa Mill) starch consisting in steeping and fruit disintegration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service