Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3275

Omega-3 and Omega-6 fatty acids in fish oil

NIST® SRM® 3275

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

fish oil

Quality Level

grade

certified reference material

form

liquid

packaging

pkg of 3 ea (2 ampule x 1.2 mL)

manufacturer/tradename

NIST®

matrix

fish

format

matrix material

shipped in

wet ice

storage temp.

2-8°C

General description

This Standard Reference Material (SRM) is intended primarily for validation of methods for determining fatty acids in fish oils and similar materials. This SRM can also be used for quality assurance when assigning values to in-house reference materials. SRM 3275 consists of three individual oils: Part 3275-1, a concentrate high in docosahexaenoic acid (DHA); Part 3275-2, an anchovy oil high in DHA and eicosapentaenoic acid (EPA); and Part 3275-3, a concentrate containing 60 % long-chain omega-3 fatty acids. A unit of SRM 3275 consists of two ampoules of each of the three oils, each ampoule containing approximately 1.2 mL of material.For more information, please refer to the COA and SDS.

SRM 3275_cert

SRM 3275_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Lipids and Fatty acids: (Z,Z,Z,Z,Z)-5,8,11,14,17-Eicosapentaenoic acid (C20:5 n-3; EPA), (Z,Z,Z,Z,Z)-5,8,11,14,17-Eicosapentaenoic acid (C20:5 n-3; EPA), (Z,Z,Z,Z,Z,)-7,10,13,16,19-Docosapentaenoic acid (C22:5; DPA), (Z,Z,Z,Z,Z,Z)-4,7,10,13,16,19-Docosahexaenoic acid (C22:6 n-3; DHA), Arachidic acid, Behenic acid, Eicosanoic acid, Erucic acid, Erucic acid, Gondoic acid, Lauric acid, Lignoceric acid, Linoleic acid, Myristic acid, Myristoleic acid, Nervonic acid, Oleic acid, Palmitic acid, Palmitoleic acid, Stearic acid, Vaccenic acid, α-Linolenic acid, γ-Linolenic acid

Matrix Group: Fish and Seafood

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service