Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3247

Ginkgo biloba extract

NIST® SRM® 3247

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 3247 contains five 2 mL ampoules, each consisting of about 1 gram of extract.

SRM 3247_cert

SRM 3247_SDS

Application

The SRM is intended to validate analytical methods for determining flavonoids, terpene lactones, andtoxic elements in Ginkgo biloba extracts and similar matrices. Additionally, it can be utilized to assign values to in-house control materials for quality assurance purposes.

Features and Benefits

  • Accompanied with NIST- certified mass fraction values for flavonoid aglycones and terpene lactones, and lead in SRM 3247.
  • Reference values for arsenic and cadmium are also expressed as mass fractions on their dry-mass basis.
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Bilobalide, Ginkgolide A, Ginkgolide B, Ginkgolide C, Ginkgolide J, Isorhamnetin, Kaempferol, Quercetin, Total Aglycones, Total Terpene Lactones
Trace Elements: Arsenic (As), Cadmium (Cd), Lead (Pb)

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service