Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

77403

Supelco

ε-Caprolactam

analytical standard

Synonym(s):

2-Oxohexamethyleneimine, Aza-2-cycloheptanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
106934
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

707 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

136-138 °C/10 mmHg (lit.)

mp

68-71 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C1CCCCCN1

InChI

1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)

InChI key

JBKVHLHDHHXQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ε-Caprolactam is used in the manufacture of polyamides. It is also used to produce nylon 6.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioremediation of ε-caprolactam from Nylon-6 waste water by use of Pseudomonas aeruginosa MCM B-407.
Kulkarni RS, et al.
Current Microbiology, 37(3), 191-194 (1998)
Examination of organic trace contamination and thermo-oxidative deterioration of ε-caprolactam by high-performance liquid chromatography.
Eppert G, et al.
Journal of Chromatography A, 508, 149-158 (1990)
P Gross
Critical reviews in toxicology, 13(3), 205-216 (1984-01-01)
In general, this literature review indicates that epsilon-caprolactam has a relatively low toxicity to humans. This low-degree of toxicity is attributable in part to its rapid elimination as demonstrated by experimental studies on animals. Human studies are mostly those based
Gun-Seok Park et al.
Journal of biotechnology, 192 Pt A, 263-264 (2014-12-03)
Pseudomonas mosselii strain SJ10 is a caprolactam-degrading bacterium belonging to the class Gammaproteobacteria, which was isolated from wastewater of the nylon 6 producing Seongseo industrial complex in Daegu, Republic of Korea. Here, we report the complete genome sequence of the
Bin Tian et al.
Pharmaceutical research, 32(3), 840-851 (2014-08-29)
To predict the temperature-composition phase diagram of solid dispersions (SDs) through theoretical approaches using cinnarizine-Soluplus(®) SD as a model system and evaluate the predicted results. A complete phase diagram of cinnarizine-Soluplus(®) SD, including the solubility curve, miscibility curve and glass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service