Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91641

Supelco

Laurolactam

analytical standard

Synonym(s):

12-Aminododecanoic acid lactam, Aza-2-cyclotridecanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO
CAS Number:
Molecular Weight:
197.32
Beilstein:
122031
MDL number:
UNSPSC Code:
12352100

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

O=C1CCCCCCCCCCCN1

InChI

1S/C12H23NO/c14-12-10-8-6-4-2-1-3-5-7-9-11-13-12/h1-11H2,(H,13,14)

InChI key

JHWNWJKBPDFINM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service