Skip to Content
MilliporeSigma
All Photos(1)

Documents

95636

Supelco

Extractables and Leachables Screening Standard for LC

certified reference material, TraceCERT®, 21 components in acetonitrile, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

quality

21 components in acetonitrile

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

50 mg/L each component (nominal concentration)

application(s)

pharmaceutical

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our complete range of extractables and leachables certified reference mixes

Application

The certified reference material (CRM) is intended to be used as a calibrant for Extractables and Leachables testing in various plastic compounds.

Certified Reference Mixtures in ExtractablesScreening of Polymeric Materials: For Container–Closure Systems and Single-UseEquipment (article in bioprocess international )

Extractables Study with LC-UV/MS - Efficient identification and quantification of unknownextractables of a single-use system using the certified reference material(CRM) mix for extractables and leachables article from AnalytixReporter issue 12


Features and Benefits

  • Produced and certified under ISO/IEC 17025 and ISO 17034 accreditation
  • Traceable to certified NIST SRMs
  • Accompanied with a comprehensive Certificate of Analysis (CoA) (up to four pages), following ISO Guide 31, with data on stability, density, homogeneity, the accuracy of concentration, uncertainty, and traceability
  • Gravimetrically prepared using high-quality qNMR-characterized starting materials and qualified precision balances to ensure minimal uncertainty
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Available in well-sealed amber glass ampules

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate)
    Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
    2-(2-Hydroxy-5-methylphenyl)benzotriazole
    ε-Caprolactam
    Dibenzylamine
    Benzoic acid
    2-Mercaptobenzothiazole
    Bisphenol A
    2-Ethylhexanoic acid
    Bis(4-chlorophenyl) sulfone
    3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
    2,6-Di-tert-butyl-4-methylphenol
    1,3-Di-tert-butylbenzene
    Oleamide
    Bis(2-ethylhexyl) phthalate
    Stearic acid
    cis-13-Docosenoamide
    Tris(3,5-di-tert-butyl-4-hydroxybenzyl) isocyanurate
    Tris(2,4-di-tert-butylphenyl)phosphate
    2,4-Di-tert-butylphenol
    Palmitic acid
    See All (21)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service