Skip to Content
MilliporeSigma

Skip To

36735

Bis(2-ethylhexyl) phthalate

PESTANAL®, analytical standard

Synonym(s):

DEHP, Phthalic acid bis(2-ethylhexyl ester), Dioctyl phthalate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
204-211-0
Beilstein/REAXYS Number:
1890696
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

General description

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
672614799480030
application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

format

neat

format

neat

format

single component solution

format

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

200

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

-

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Bis(2-ethylhexyl) phthalate


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

392.0 °F - Pensky-Martens closed cup

flash_point_c

200 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jian Hou et al.
Ecotoxicology and environmental safety, 182, 109384-109384 (2019-07-05)
Exposure to polycyclic aromatic hydrocarbons (PAHs) and phthalates link to oxidative stress and inflammatory response, which exert cellular aging. However, modification effect of seasonal factor on the association of PAHs or phthalates exposure with relative telomere length (RTL) or mitochondrial
Alessia Arena et al.
Food chemistry, 396, 133721-133721 (2022-07-24)
Phthalic acid esters (PAEs), known as phthalates, have a variety of industrial applications, mainly related to their ability to increase plastic softness,flexibility, and durability. With regard to toxicological aspects, several studies suggest that phthalates would act as endocrine-disrupting chemicals, and
Anne-Marie Saillenfait et al.
Journal of applied toxicology : JAT, 33(9), 1027-1035 (2013-06-12)
In utero exposure to the phthalate ester plasticizer di-n-hexyl phthalate (DnHP) is known to affect the development of the male reproductive system and induce alterations in androgen-dependent tissues of male rat offspring. Male reproductive malformations produced by several phthalates have



Global Trade Item Number

SKUGTIN
36735-1G04061838082404

Questions

1–5 of 5 Questions  
  1. What is the concentration of this product?

    1 answer
    1. This product is a neat or pure liquid that has not been diluted in a solvent. This material has a molecular weight of 390.56, a density of 0.985 g/mL, and a minimum purity of 98%. The molarity may range from 2.47 M to 2.52M.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Hello I would like to know the density of this product (pestanal Bis(2-ethylhexyl) phthalate; CAS Number: 117-81-7). I searched but couldn't find it. I would be very grateful if you could help me. Thank you from now.

    1 answer
    1. The density of this item is 0.985 g/mL at 20 °C (lit.)

      Helpful?

  5. Good morning! What is the solubilty of the substance DEHP, please? And the solubility of the benzyl butyl phthalate? Thank you!

    1 answer
    1. Both compounds are only slightly soluble in water. Product 36735 is not tested for solubility. According to the literature, it is soluble in ethanol, DMSO, benzene, and ether. No rate is reported. Also, according to the literature, the solubility of benzyl butyl phthalate (CAS 85-68-7,) is in chloroform, DMSO, and methanol. No rate is reported.

      Helpful?

Reviews

No rating value

Active Filters