Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02800

Sigma-Aldrich

ε-Caprolactam

for synthesis

Synonym(s):

ε-Caprolactam, 6-Aminocaproic acid lactam, 2-Oxohexamethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-313-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.0014 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

scales

autoignition temp.

395 °C (DIN 51794)

potency

1660 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-8 % (v/v)

pH

7.0-8.5 (20 °C, 333 g/L in H2O)

bp

270.8 °C/1013 hPa

mp

68-70 °C

transition temp

flash point 152 °C (DIN 51758)

solubility

4560 g/L

density

1.01 g/cm3 at 80 °C

bulk density

500‑550 kg/m3

storage temp.

2-30°C

SMILES string

N1CCCCCC1=O

InChI

1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)

InChI key

JBKVHLHDHHXQEQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 71 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service