Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM01483

Supelco

ε-Caprolactam

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

CAP, 2-Oxohexamethyleneimine, Aza-2-cycloheptanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
106934
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

product line

TraceCERT®

form

crystals

autoignition temp.

707 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

bp

136-138 °C/10 mmHg (lit.)

mp

68-71 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCCCCN1

InChI

1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)

InChI key

JBKVHLHDHHXQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Other Notes

Ion Mobility: TWCCSN2 value of 121.9 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N CRM01483 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Ascentis® C18 column facilitates LC-UV/MS analysis of extractables from single-use systems, ensuring reliable identification and quantification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service