Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.06454

Supelco

Dichloromethane

hypergrade for organic trace analysis SupraSolv®

Synonym(s):

DCM, Methylene chloride, Methylene dichloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-838-9
NACRES:
NA.03
technique(s):
GC/MS: suitable
application(s):
general analytical
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

SupraSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

1223 °F
605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

22 %

technique(s)

GC/MS: suitable

dilution

( suitable for gas chromatography (GC))

impurities

≤0.005% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

solubility

20 g/L

density

1.325 g/mL at 25 °C (lit.)

application(s)

general analytical

storage temp.

2-30°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® hypergrade for organic trace analysis is a unique and universal solvent quality which can be employed for all three GC detection methods (ECD, FID and MS). Thanks to the large retention time range, even low-boiling substances can be reliably analyzed.

Application


  • Evaluation of the antifungal photodynamic activity of Thymophylla pentachaeta extracts against Candida albicans and its virulence factors.: This study explores the antifungal properties of Thymophylla pentachaeta, highlighting the use of dichloromethane in the extraction process, which is critical for isolating active compounds with potential therapeutic applications (Cordisco et al., 2021).


  • Amazonian medicinal smokes: Chemical analysis of Burseraceae pitch (breu) oleoresin smokes and insights into their use on headache.: This research details the chemical components of Burseraceae pitch smokes, with a focus on compounds extracted using dichloromethane. These findings provide insights into traditional medicine and its applications in treating headaches (da Cruz Albino et al., 2021).


  • Chemical characterization, antioxidant and enzyme inhibitory effects of Mitracarpus hirtus extracts.: This paper discusses the antioxidant and enzyme inhibitory properties of Mitracarpus hirtus, where dichloromethane was used in the extraction process to isolate bioactive compounds, emphasizing its utility in pharmacological screenings (Etienne et al., 2021).


  • Assessment of in vitro anthelmintic activity and bio-guided chemical analysis of BRS Boyrá pineapple leaf extracts.: Investigates the anthelmintic activities of pineapple leaf extracts, using dichloromethane for the extraction and isolation of active compounds, providing valuable insights into the potential veterinary applications of these extracts (Rodrigues et al., 2020).


  • Head Louse Feces: Chemical Analysis and Behavioral Activity.: This study analyzes the chemical composition of head louse feces, including the application of dichloromethane in the extraction process. The findings offer a deeper understanding of the behavioral responses associated with louse infestations (Galassi et al., 2020).



Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 3.0 mg/l
Water: ≤ 0.005 %
Colour: ≤ 10 Hazen
GC/ECD (retention range 1,2,4-tri-chlorobenzene to decachlorobiphenyle,individual signals (lindane standard)): ≤ 2 pg/ml
GC/FID (retention range n-undecane ton-tetracontane,individual signals (n-tetradecane standard)): ≤ 2 ng/ml
GC/MSD (retention range n-undecane ton-tetracontane, scaning area 30 - 600 amu,individual signals (n- tetradecane standard)): ≤ 2 ng/ml
Recommended for analysis of polychlorinated Dibenzodioxins and polychlorinated Dibenzofurans (PCDD/F).
The product is stabilized with 2-Methyl-2-buten.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service