Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
n-Hexane, hypergrade for organic trace analysis SupraSolv®
vapor density
~3 (vs air)
Quality Segment
vapor pressure
256 mmHg ( 37.7 °C), 5.2 psi ( 37.7 °C), ~132 mmHg ( 20 °C)
product line
SupraSolv®
assay
≥99% (GC)
form
liquid
autoignition temp.
453 °F
potency
25000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)
expl. lim.
7.7 %
technique(s)
GC/MS: suitable, gas chromatography (GC): suitable
impurities
≤0.005% Water, ≥99.9% Sum of hexane isomers + methylcyclopentane (GC)
evapn. residue
≤3.0 mg/L
color
APHA: ≤10
refractive index
n20/D 1.375 (lit.)
kinematic viscosity
0.50 cSt(20 °C)
bp
69 °C (lit.)
mp
−95 °C (lit.)
transition temp
flash point -22 °C
solubility
0.0095 g/L
density
0.659 g/mL at 25 °C (lit.)
suitability
suitable for residue analysis
storage temp.
2-30°C
SMILES string
CCCCCC
InChI
1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI key
VLKZOEOYAKHREP-UHFFFAOYSA-N
General description
Application
Features and Benefits
- Universal detector compatibility, eliminating the need for detector-specific solvent selection and simplifying laboratory workflows
- Extended and broader specified retention time range, enabling reliable detection of both low- and high-boiling analytes within a single analytical run
- Batch-to-batch consistency and rigorous lot-specific quality control, ensuring reproducibility across analytical series
- Minimized matrix-induced interferences, critical for achieving accurate quantification at ppt to ppb concentration levels
Analysis Note
- ≤ 1 ng/mL interfering impurities relative to tetrachloromethane by GC-ECD
- ≤ 2 pg/mL for lindane equivalents by GC-ECD
- ≤ 2 ng/mL for n-tetradecane equivalents by GC-FID and GC-MSThese specifications ensure that solvent-derived background signals remain below the detection threshold of the most sensitive analytical methods, thereby safeguarding the integrity of trace-level quantification.
Other Notes
Legal Information
Still not finding the right product?
Explore all of our products under n-Hexane
Signal Word
Danger
Precautionary Statements
Target Organs
Central nervous system, Nervous system
Storage Class
3 - Flammable liquids
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3
Hazard Codes
Flash Point (°F)
-7.6 °F - closed cup
Flash Point (°C)
-22 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



