Skip to Content
MilliporeSigma

Skip To

1.00668

Dichloromethane

for gas chromatography MS SupraSolv®

Synonym(s):

DCM, Methylene dichloride, Methylene chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH2Cl2
CAS Number:
Molecular Weight:
84.93
UNSPSC Code:
12352101
NACRES:
NA.03
EC Index Number:
200-838-9
MDL number:
Beilstein/REAXYS Number:
1730800
Assay:
≥99.8% (GC)
Technique(s):
GC/MS: suitable
Bp:
39.8-40 °C (lit.)
Vapor pressure:
24.45 psi ( 55 °C), 6.83 psi ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C), 6.83 psi ( 20 °C)

product line

SupraSolv®

assay

≥99.8% (GC)

form

liquid

autoignition temp.

1223 °F, 605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

expl. lim.

22 %

technique(s)

GC/MS: suitable

impurities

≤0.01% Water

evapn. residue

≤5.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C, −95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

General description

SupraSolv® MS is a dedicated high-purity solvent grade specifically engineered for sample preparation and extraction in gas chromatography coupled with mass spectrometric detection (GC-MS)

Application

SupraSolv® MS solvents are routinely employed in highly demanding GC-MS-based analytical workflows, including polychlorinated biphenyl (PCB) determination in water and soil, pesticide residue analysis in food commodities such as fruits and vegetables, environmental contaminant profiling, dioxin screening, and multi-residue methods — applications in which unambiguous mass spectral identification and accurate trace-level quantification are of paramount regulatory and scientific importance.

Analysis Note

SupraSolv® MS solvents are produced via purification processes, yielding:
  • Extremely low levels of non-volatile residues and evaporation residues, preventing column contamination and ion source fouling
  • Minimal intrinsic volatile organic impurities, verified by GC-MS testing across the full chromatographic retention time window

Other Notes

Complete GC Workflow Guide for ICH Q3C(R9) and USP<467>Compliance

Explore various labsafety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Dichloromethane


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I don't see the type of packaging listed. Is this product available in a glass bottle?

    1 answer
    1. Yes, this product is packaged in glass bottles.

      Helpful?

Reviews

No rating value

Active Filters