Skip to Content
MilliporeSigma
All Photos(1)

Documents

DX0831

Supelco

Dichloromethane

OmniSolv®

Synonym(s):

Methylene Chloride, FM approval, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-838-9

Agency

suitable for EPA 1613

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.005% Water (H2O)
≤0.2 μeq/g Titrable acid
≤2 ng/L Capillary ECD responsive substances (as PCNB)
≤3 μg/L Capillary FID responsive substances (as decane)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

UV cutoff

≤231 nm

UV absorption

λ: 231 nm Amax: ≤1.00
λ: 235 nm Amax: ≤0.40
λ: 240 nm Amax: ≤0.20
λ: 250 nm Amax: ≤0.01
λ: 260 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane, also known as Methylene chloride, is commonly used as a solvent with a wide range of applications. Due to high solubility, it is used to perform nonlinear transmission (NLT) measurements. Additionally, it is used as an effective extraction solvent, allowing for the separation and purification of desired substances from mixtures.

Application

Dichloromethane is used as a solvent:
  • To prepare spectroscopic samples.
  • For the dilution of s-diphenylaminostilbene which is used as a reference material for two-photon excited fluorescence (2PEF) measurement.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minCapillary ECD responsive substances (as PCNB)2ng/L maxCapillary FID responsive substances (as decane)3µg/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)500ppt maxFormClear liquidFree halogensTo pass testIdentity (IR-spectrum)ConformsRefractive index (at 20°C)Titrable acid0.2µeq/g maxUV Abs. at 231 nm1.00AU maxUV Abs. at 235 nm0.40AU maxUV Abs. at 240 nm0.20AU maxUV Abs. at 250 nm0.01AU maxUV Abs. at 260 nm0.005AU maxUV Cut-off231nm maxWater (H2O)0.005% max
Evaporation Number: 1.9

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrolo [3, 2-b] pyrroles?From Unprecedented Solvatofluorochromism to Two-Photon Absorption.
Daniel F H, et al.
Chemistry?A European Journal , 21, 18364-18374 (2015)
Gelation and swelling behavior of end-linked hydrogels prepared from linear poly (ethylene glycol) and poly (amidoamine) dendrimers
Burcu U,et.al
Polymer, 47, 8173-8182 (2006)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service