If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
1.03654
n-Heptane
hypergrade for LC-MS LiChrosolv®
Synonym(s):
Heptane
About This Item
83 mmHg ( 37.7 °C)
Recommended Products
vapor density
3.5 (vs air)
Quality Level
vapor pressure
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
product line
LiChrosolv®
Assay
≥99.3% (GC)
form
liquid
autoignition temp.
433 °F
expl. lim.
7 %
greener alternative product characteristics
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
LC/MS: suitable
impurities
≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water
≤10 ppb Al (Aluminium)
≤10 ppb Ca (Calcium)
≤10 ppb Fe (Iron)
≤10 ppb Mg (Magnesium)
≤10 ppb Na (Sodium)
≤5 ppb K (Potassium)
evapn. residue
≤2.0 mg/L
transmittance
210 nm, ≥50%
220 nm, ≥80%
245 nm, ≥98%
refractive index
n20/D 1.387 (lit.)
bp
98 °C (lit.)
mp
−91 °C (lit.)
transition temp
flash point -4 °C
solubility
0.05 g/L
density
0.684 g/mL at 25 °C (lit.)
greener alternative category
storage temp.
2-30°C
SMILES string
CCCCCCC
InChI
1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3
InChI key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Pt(PPh(3))(4) and Pt(PPh(3))(4)@IL catalyzed hydroboration of ketones.: This study explores the use of n-Heptane as a solvent in the hydroboration of ketones catalyzed by Pt(PPh(3))(4) and Pt(PPh(3))(4)@IL, demonstrating its application in organometallic chemistry and catalytic processes (Krupa et al., 2023).
- Imidazolium-based ionic liquids as dispersants to improve the stability of asphaltene in Egyptian heavy crude oil.: This research uses n-Heptane in the analysis of asphaltene stability, highlighting its role in petroleum chemistry and the development of more efficient dispersants (Ghanem et al., 2023).
- A countercurrent chromatography solvent system based on deep eutectic solvents for separation of cis- and trans-crocetin from Gardenia jasminoides Ellis.: This study employs n-Heptane in a novel solvent system for countercurrent chromatography, showcasing its utility in natural product isolation and analytical chemistry (Wen et al., 2023).
- Volatile Organic Compounds in Disposable Diapers and Baby Wipes in the US: A Survey of Products and Health Risks.: The study includes n-Heptane in the analytical methods to detect volatile organic compounds, emphasizing its importance in environmental health and safety assessments (Lin et al., 2023).
- Countercurrent chromatography separation of vitamin E isomers in a co-current mode.: This research uses n-Heptane in countercurrent chromatography to separate vitamin E isomers, demonstrating its application in nutritional chemistry and quality control (Hu et al., 2023).
Preparation Note
Analysis Note
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.005 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Al (Aluminium): ≤ 10 ppb
Ca (Calcium): ≤ 10 ppb
Fe (Iron): ≤ 10 ppb
K (Potassium): ≤ 5 ppb
Mg (Magnesium): ≤ 10 ppb
Na (Sodium): ≤ 10 ppb
Transmission (at 210 nm): ≥ 50 %
Transmission (at 220 nm): ≥ 80 %
Transmission (from 245 nm): ≥ 98 %
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 2 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI negative): ≤ 20 ppb
Filtered by 0.2 μm filter.
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
24.8 °F
Flash Point(C)
-4 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service