L070150
Fast Deprotection Cap 2
45-55 g/L (TAC2O content, gravimetric)
Synonym(s):
Fast Deprotection Cap A
About This Item
Recommended Products
type
for DNA synthesis
for DNA synthesis
Quality Level
product line
Proligo Reagents
form
liquid
storage condition
dry at room temperature
concentration
45-55 g/L (TAC2O content, gravimetric)
technique(s)
DNA synthesis: suitable
color
clear
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Preparation Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
35.6 °F - (Acetonitrile)
Flash Point(C)
2.0 °C - (Acetonitrile)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service