Skip to Content
MilliporeSigma
All Photos(1)

Documents

L032100

Sigma-Aldrich

DCI Activator

0.5 M

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

58-64 g/L (DCI content, titration)

impurities

≤30 ppm water content (Karl Fischer)
≤30 ppm water content (Karl Fischer)

color

clear
clear

Looking for similar products? Visit Product Comparison Guide

General description

Activator solution, containing powerful activators like 4,5-dicyanoimidazole(DCI), 5-(3,5-bis(trifluoromethyl)phenyl)-1H-tetrazole (Activator 42) or5-(Ethylthio)-1H-tetrazole (ETT) in acetonitrile, are mixed with solutions ofphosphoramidites during the extension step. The activator reacts with theamidite group to form a highly reactive intermediate. The intermediate thenforms an internucleotide bond with the detritylated 5′hydroxyl group of thegrowing oligonucleotide chain.

Preparation Note

Dicycanoimidazole /Acetonitrile 6/100 (w/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service