Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

555339

Sigma-Aldrich

Cap Mix A

80% Tetrahydrofuran, 10% Acetic Anhydride, 10% Pyridine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005
NACRES:
NA.21
bp:
65 °C (lit.)
Pricing and availability is not currently available.

Product Name

Cap Mix A, (80% Tetrahydrofuran, 10% Acetic Anhydride, 10% Pyridine),

impurities

≤0.01% water
water

Quality Level

bp

65 °C (lit.)

density

0.916 g/mL at 25 °C (lit.)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service