Skip to Content
MilliporeSigma
Fast Deprotection Cap 2 45-55 g/L (TAC2O content, gravimetric)

Fast Deprotection Cap 2

Fast Deprotection Cap 2 45-55 g/L (TAC2O content, gravimetric)
Synonym(s):
Fast Deprotection Cap A

Products

Product Category
Brand
Color
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing