Skip to Content
MilliporeSigma
All Photos(1)

Documents

L040030

Sigma-Aldrich

Cap A

configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

shelf life

365 days

concentration

97.0-119.0 g/L (Acetic Anhydride content, titration)

color

clear colorless, to brownish

compatibility

configured for ABI

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Cap A solution contains acetic anhydrideanhydride, lutidine and tetrahydrofuran (THF). It is used after the phosphoramidite reaction/coupling step. The solution is configured for ABI.

Biochem/physiol Actions

Cap A solution aborts chains bearing unreacted 5′ hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′ hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Preparation Note

Tetrahydrofurane / Lutidine / Acetic Anhydride 8/1/1 (v/v/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service