Key Documents
CLS88253424
Pyrex® bushing-type reducing adapter
female joint: ST/NS 34/45, male joint: ST/NS 24/40
Synonym(s):
glass adapter, glass adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
female joint: ST/NS34/45
male joint: ST/NS24/40
manufacturer/tradename
Corning 8825-3424
Corning 8825
availability
not available in EU
L
59 mm
Related Categories
General description
Heavy walls for mechanical strength and heavy upper rim for good finger grip. Shorter than conventional type for more compact assemblies. Useful for attaching jointed thermometers.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)