Key Documents
CLS88202429
Pyrex® enlarging adapter
female joint: ST/NS 24/40, male joint: ST/NS 29/42
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41121815
Joint
female joint: ST/NS24/40
male joint: ST/NS29/42
manufacturer/tradename
Corning 8820-2429
availability
not available in EU
L
85 mm
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)