Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A549 Cell Line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86012804, human lung (carcinoma), Epithelial

Synonym(s):

A-549 Cells, A549 Cells, NCI-A549 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human lung (carcinoma)

packaging

tube of 5 μg 86012804-DNA-5UG
pkg of vial of cells 86012804-1VL

growth mode

Adherent

karyotype

Hypotriploid

morphology

Epithelial

products

Lecithin, High amounts of de-saturated fatty acids.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung carcinoma

Cell Line Description

Derived from a 58 year old Caucasian male. The cells can synthesise lecithin utilising the cytidine diphosphocholine pathway. Occasional cells may also contain inclusion bodies although they are not known to carry any human pathogen.

Application

Enzymatic studies
Human A549 cell line has been used:
  • to study the ability of 2-(2,4-dihydroxyphenyl)thieno-1,3-thiazin-4-one (BChTT) to inhibit cancer cell proliferation and identify mechanisms involved in its actions[1]
  • to study the effects of insulin and insulin-like growth factor 1 (IGF1) on proliferation and apoptosis in A549 cells[2]
  • to culture A549 cells in different media over a long term and identify conditions for promoting alveolar type II differentiation characteristics in A549 cells[3]

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 11
D16S539: 11,12
D5S818: 11
D7S820: 8,11
THO1: 8,9.3
TPOX: 8,11
vWA: 14

Culture Medium

Ham′s F12K + 2mM Glutamine + 10% Foetal Bovine Serum FBS / FCS.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% Trypsin or tTrypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

Learn how to quantify spheroid formation, roundness, and circularity. Discover Millicell® Ultra-low Attachment plates, an ideal 3D cell culture model for uniform spheroid formation.

Learn about the similarities and differences in spheroid formation, roundness, and circularity between Millicell® ultra-low attachment plates and other ULA plates.

See All

Related Content

Millicell® Ultra-low Attachment plates facilitate 3D cell culture with detailed application information for cell seeding to analysis.

Millicell® Ultra-low Attachment plates facilitate 3D cell culture with detailed application information for cell seeding to analysis.

Questions

1–9 of 9 Questions  
  1. What HLA-ABC phenotype do A549 cells show?

    1 answer
    1. Please view the article in the link below for more information regarding the phenotype of this cell line:
      https://www.culturecollections.org.uk/media/126668/jim-plos-one-article-a549.pdf

      Please visit the ECACC website for additional documentation and information regarding this cell line:
      https://www.culturecollections.org.uk/products/celllines/generalcell/detail.jsp?refId=86012804&collection=ecacc_gc

      Helpful?

  2. What is cell line?

    1 answer
    1. Hello. Human A549 cell line is a human lung carcinoma that was derived from a 58 year old Caucasian male.

      Helpful?

  3. What medium should I use when I thaw the Cell Line cells?

    1 answer
    1. The medium for each cell line is listed on the product page. It is specific to each cell line. If not visible on the Sigma-Aldrich website product page, the HPA website also contains the same information (www.hpacultures.org.uk). The product number for the cell line is the same on either site.

      Helpful?

  4. Are my Cell Line cells mycoplasma tested?

    1 answer
    1. ECACC routinely tests all manufactured cell banks for mycoplasma.

      Helpful?

  5. How many Cell Line cells are in the vial?

    1 answer
    1. Each vial of cells contains 2-3 x 106 cells in 1 ml of freezing media. This is in a 1.8 mL ampule.  For exceptionally large cells, counts may decrease. Suspension cells, generally smaller cells, may contain as many as 4-5 x 106 cells/vial to assure optimal viability upon thaw.

      Helpful?

  6. What passage are my Cell Line cells?

    1 answer
    1. If the passage of the cells is known, it is listed on the product page at the HPA website (www.hpacultures.org.uk).

      Helpful?

  7. Why is Vapor Phase Liquid Nitrogen preferred for storage of Cell Line cells?

    1 answer
    1. If ampules are immersed into liquid phase of liquid nitrogen, it increases the risk of the liquid seeping into the vial. This could lead to problems of cross-contamination and increased risk of the ampule exploding when thawed.

      Helpful?

  8. How do I handle Cell Line frozen cells upon arrival?

    1 answer
    1. Upon receipt, frozen ampules should be transferred directly to vapor phase liquid nitrogen without delay (-135°C) or liquid phase liquid nitrogen if vapor phase is not available. DO NOT use a -80°C freezer as an alternative; this will result in loss of viability.

      Helpful?

  9. Are the Cell Line cells tested for viral pathogens?

    1 answer
    1. HPA Cultures does not perform any viral testing on the cell lines.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service