Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A375-C5 Cell Line human

97011320, human skin, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

A375-C5 Cell Line human, 97011320

biological source

human skin

description

Human malignant melanoma IL-1 insensitive

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human malignant melanoma IL-1 insensitive

Cell Line Description

The cell line A375-C5 is unresponsive to the anti-proliferative effect of IL-1. It has been derived from the parental line A373 (Sigma catalogue no. 88113005) which was subjected to cloning by limiting dilution. This method resulted in the establishment of A375-C5 and an IL-1 sensitive clone, A375-C6 (Sigma catalogue no. 97011321). The unresponsiveness of A375-C5 is probably due to an IL-1 receptor deficiency. The cells have been eradicated from mycoplasma at ECACC.

Application

IL-1 resistance studies, receptor studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service