Recommended Products
biological source
human cervix
Quality Level
form
liquid
growth mode
Adherent
karyotype
2n = 46
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human cervix carcinoma
Cell Line Description
These cells were originally Chang conjunctival cells adapted by Wong and Kilbourne, and designated as conjunctival ′D′ cells. Susceptible to swine influenza viruses, supports production of plaque formation by a variety of influenza virus subtypes. Has HeLa markers, exhibits type A G6PD. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Epithelial with occasional giant cells. Ethnicity: Black.
Application
Plaque assays, cloning, genetic and virus studies: swine influenza viruses
Culture Medium
Medium 199 (HBSS) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2using EDTA or 0.25% trypsin; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
In vitro, 14(5), 469-475 (1978-05-01)
Seed stocks of human cell lines deposited at the American Type Culture Collection (ATCC) have been examined for cross-contamination with HeLa cells using Giemsabanded marker chromosomes. Sixteen additional cell lines investigated have been found to exhibit marker chromosomes typical of
Continuous subcultivation of epithelial-like cells from normal human tissues.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 87(2), 440-443 (1954-11-01)
Examination of ATCC stocks for HeLa marker chromosomes in human cell lines.
Nature, 259(5540), 211-213 (1976-01-22)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service