Documentos
Z302864
Quickfit® round-bottom flasks
capacity 2,000 mL, joint: ST/NS 29/32
Sinónimos:
FR2L/4S
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
short
single-neck
joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Quickfit FR2L/4S
availability
not available in USA and Canada
capacity
2,000 mL
General description
Short neck with ground-glass joint.
Legal Information
Quickfit is a registered trademark of SciLabware Limited