Documentos clave
Z168904
Sigma-Aldrich
Precision crucible tongs
L 9 1/2 in. (241 mm)
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
material
stainless steel
L
9 1/2 in. (241 mm)
General description
Stainless steel tongs with serrated tapered tips, which meet precisely when closed.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)