Key Documents
BR452210
BRAND® Inoculation loop with needle, disposable
volume 10 μL, blue
Synonym(s):
Inoculation loops
Select a Size
1000 EA
$134.00
$134.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1000 EA
$134.00
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
$134.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
blue
polystyrene
sterility
sterile; γ-irradiated
packaging
pack of 1000 ea
pkg of 1000 ea (50 bags of 20)
manufacturer/tradename
BRAND 452210
volume
10 μL
Related Categories
General description
For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface. γ-ray sterilized.
Features and Benefits
  • Sterile (SAL 10-6)
  • High flexibility for agar-saving smears
  • Usable at both ends
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)