Key Documents
BR452215
BRAND® Inoculation loop with needle, disposable
volume 1+10 μL, yellow
Synonym(s):
Inoculation loops
Select a Size
1000 EA
$138.00
$138.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1000 EA
$138.00
About This Item
UNSPSC Code:
42142523
NACRES:
NB.22
$138.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polystyrene
yellow
sterility
sterile; γ-irradiated
packaging
pack of 1000 ea
pkg of 8000 ea (400 bags of 20)
manufacturer/tradename
BRAND 452215
volume
1+10 μL
Related Categories
General description
For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface. γ-ray sterilized.
Features and Benefits
  • Sterile (SAL 10-6)
  • High flexibility for agar-saving smears
  • Usable at both ends
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)