Skip to Content
Merck
All Photos(1)

Key Documents

I8263

Inoculating Loop

loop size 1 μL

Synonym(s):

inoculating loops, inoculation loop, inoculation loops

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$306.00

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
500 EA
$306.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polystyrene

sterility

sterile; γ-irradiated

loop size

1 μL

color

color-coded blue

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Loops are smooth and flexible to facilitate uniform streaking without damaging the gel surface. Disposable inoculating loops do not require flaming and thus eliminate the risk of infection. Loops are not calibrated.

Packaging

In tray packs containing 10 loops

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service