Skip to Content
Merck
All Photos(1)

Key Documents

I7898

Sigma-Aldrich

Nunc® inoculating loops

blue, pkg of 600 (50 sleeves of 12 each), size 10 μL

Synonym(s):

inoculating loop, inoculation loop, inoculation loops

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$353.00

$353.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
1 PKG
$353.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

$353.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


material

polystyrene

sterility

sterile

packaging

pkg of 600 (50 sleeves of 12 each)

manufacturer/tradename

Nunc 254437

size

10 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

  • Accurate to semiquantitative standard for handling samples
  • Surface treated to enhance adhesion of the droplets
  • Reduced risk of spreading microorganisms since the loops are disposable
  • Non toxic

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service