Key Documents
HS81121C
Inoculating Needle/Loops
loop volume 10 μL,
flexible
, sterile, blue
Synonym(s):
Inoculation Tools, Sterile Inoculating Loops
Select a Size
1000 EACH
$123.00
$123.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1000 EACH
$123.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$123.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
blue
sterility
sterile
packaging
pack of 1000 ea
manufacturer/tradename
Heathrow Scientific HS81121C
loop volume
10 μL
General description
The Inoculating Needle/Loops combination provides choice to spread or pick colonies and cultures. Transfer bacterial cultures without risk of contamination.
Features and Benefits
  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Constructed of flexible polypropylene
  • Sterile, 20 per zip bag with lot-to-lot traceability
  • Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)