Skip to Content
Merck
All Photos(1)

Documents

361437

Sigma-Aldrich

cis-1,2-Cyclopentanediol

98%

Synonym(s):

cis -1,2-Dihydroxycyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8(OH)2
CAS Number:
Molecular Weight:
102.13
Beilstein:
1901117
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

bp

108-109 °C/20 mmHg (lit.)

mp

24-28 °C (lit.)

SMILES string

O[C@@H]1CCC[C@@H]1O

InChI

1S/C5H10O2/c6-4-2-1-3-5(4)7/h4-7H,1-3H2/t4-,5+

InChI key

VCVOSERVUCJNPR-SYDPRGILSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanting Ni et al.
Bioorganic & medicinal chemistry letters, 18(5), 1567-1572 (2008-02-12)
Bacteria can coordinate community-wide behaviors through quorum sensing, that is, the secretion and sensing of autoinducer (AI) molecules. Bacterial quorum sensing is implicated in the regulation of pathologically relevant events such as biofilm formation, bacterial virulence, and drug resistance. Inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service