Merck
  • Pyrogallol and its analogs can antagonize bacterial quorum sensing in Vibrio harveyi.
Pyrogallol and its analogs can antagonize bacterial quorum sensing in Vibrio harveyi.
Bioorganic & medicinal chemistry letters (2008-02-12)
Nanting Ni, Gaurav Choudhary, Minyong Li, Binghe Wang
ABSTRACT
Bacteria can coordinate community-wide behaviors through quorum sensing, that is, the secretion and sensing of autoinducer (AI) molecules. Bacterial quorum sensing is implicated in the regulation of pathologically relevant events such as biofilm formation, bacterial virulence, and drug resistance. Inhibitors of bacterial quorum sensing could therefore be useful therapeutics. Herein we report for the first time the discovery of several pyrogallol compounds as single digit micromolar inhibitors of bacterial quorum sensing in Vibrio harveyi.
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
7,8-Dihydroxycoumarin, ≥97%, powder
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
cis-1,2-Cyclopentanediol, 98%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
2,3-Dihydroxypyridine, 95%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
2,3,4-Trihydroxybenzaldehyde, 98%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)