Skip to Content
Merck
All Photos(1)

Key Documents

192805

Sigma-Aldrich

1,3-Cyclopentanediol, mixture of cis and trans

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$406.00

$406.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$406.00

About This Item

Linear Formula:
C5H8(OH)2
CAS Number:
Molecular Weight:
102.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$406.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

80-85 °C/0.1 mmHg (lit.)

mp

40 °C (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

OC1CCC(O)C1

InChI

1S/C5H10O2/c6-4-1-2-5(7)3-4/h4-7H,1-3H2

InChI key

NUUPJBRGQCEZSI-UHFFFAOYSA-N

Application

1,3-Cyclopentanediol was used in the synthesis of 1,3-cyclopentanediol bis(4-methylbenzenesulfonate)[1] and β-hydroxy-substituted cyclic carbonyl compounds[2]. It was also used in direct enantiomer resolution of diols without derivatization on a chiral polysiloxane by capillary gas chromatography[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct asymmetric a-hydroxylation of 2-hydroxymethyl ketones.
Paju A, et al.
Tetrahedron, 58(36), 7321-7326 (2002)
Direct resolution of enantiomeric diols by capillary gas chromatography on a chiral polysiloxane derived from (R, R)-tartramide.
Nakamura K, et al.
Analytical Chemistry, 62(5), 539-541 (1990)
Stepwise Introduction of p-Electron Cross-Conjugation: A Possible Access to [5] Radialenes?
Geneste F, et al.
The Journal of Organic Chemistry, 62(16), 5339-5343 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service