Skip to Content
Merck
All Photos(1)

Key Documents

797081

Sigma-Aldrich

3-Cyclopenten-1-ol

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$274.00

$274.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$274.00

About This Item

Empirical Formula (Hill Notation):
C5H8O
Molecular Weight:
84.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$274.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

functional group

hydroxyl

SMILES string

OC1CC=CC1

InChI

1S/C5H8O/c6-5-3-1-2-4-5/h1-2,5-6H,3-4H2

InChI key

WEIMJSIRDZDHAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.4 °F

Flash Point(C)

41.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service