Skip to Content
Merck
All Photos(2)

Key Documents

179035

Sigma-Aldrich

cis-1,5-Cyclooctanediol

98%

Synonym(s):

cis-1,5-Cyclooctanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14(OH)2
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

bp

105-107 °C/0.05 mmHg (lit.)

mp

73-75 °C (lit.)

functional group

hydroxyl

SMILES string

O[C@H]1CCC[C@@H](O)CCC1

InChI

1S/C8H16O2/c9-7-3-1-4-8(10)6-2-5-7/h7-10H,1-6H2/t7-,8+

InChI key

BDNXUVOJBGHQFD-OCAPTIKFSA-N

Application

cis-1,5-Cyclooctanediol was used in the synthesis of cis-1,5-diaminocyclooctane[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John C Poutsma et al.
Chemical communications (Cambridge, England), (16)(16), 2040-2041 (2003-08-26)
The gas phase basicity of the title compound has been determined to be greater than that of putrescine, making it the most basic primary diamine measured to date.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service